Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024852.3 | NP_079128.2 | 9, 10 | 1487 | ||
NM_177422.2 | NP_803171.1 | 7, 8 | 1229 | |||
XM_005270575.3 | XP_005270632.1 | 10, 11 | 6941 | |||
XM_005270576.3 | XP_005270633.1 | 4, 5 | 1725 | |||
XM_011540879.2 | XP_011539181.1 | 9, 10 | 2369 | |||
XM_011540882.2 | XP_011539184.1 | 6, 7 | 584 | |||
XM_017000523.1 | XP_016856012.1 | 10, 11 | 6957 | |||
XM_017000524.1 | XP_016856013.1 | 5, 6 | 755 | |||
XM_017000525.1 | XP_016856014.1 | 6, 7 | 589 | |||
XM_017000526.1 | XP_016856015.1 | 5, 6 | 548 | |||
XM_017000527.1 | XP_016856016.1 | 5, 6 | 727 | |||
XM_017000528.1 | XP_016856017.1 | 6, 7 | 676 | |||
GenBank mRNA | AK022827.1 | - | 9, 10 | 1242 | ||
AK292870.1 | - | 9, 10 | 1453 | |||
AK302289.1 | - | 2, 3 | 236 | |||
AK303929.1 | - | 6, 7 | 771 | |||
BC025769.1 | - | 7, 8 | 1053 |