Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHODL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204174.1 | NP_001191103.1 | 6 | 1957 | ||
NM_001204175.1 | NP_001191104.1 | 6 | 1923 | |||
NM_001204176.1 | NP_001191105.1 | 7 | 2023 | |||
NM_001204177.1 | NP_001191106.1 | 5 | 1820 | |||
NM_001204178.1 | NP_001191107.1 | 6 | 1920 | |||
NM_024944.2 | NP_079220.2 | 6 | 2332 | |||
XM_011529453.2 | XP_011527755.1 | 5 | 2463 | |||
XM_011529457.2 | XP_011527759.1 | 7 | 2329 | |||
XM_017028273.1 | XP_016883762.1 | 5 | 1849 | |||
GenBank mRNA | AF257472.1 | - | 6 | 2332 | ||
AK022689.1 | - | 7 | 2205 | |||
AY358608.1 | - | 6 | 2318 |