Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025077.3 | NP_079353.3 | 4 | 869 | ||
XM_005270412.3 | XP_005270469.1 | 4 | 852 | |||
XM_005270413.4 | XP_005270470.1 | 4 | 1129 | |||
GenBank mRNA | AK024011.1 | - | 4 | 390 | ||
AK093320.1 | - | 4 | 556 | |||
AK298162.1 | - | 4 | 345 | |||
AY169960.1 | - | 4 | 866 | |||
BC009364.2 | - | 4 | 303 | 4302712 | ||
CR457320.1 | - | 4 | 286 |