Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025077.3 | NP_079353.3 | 7 | 1424 | ||
XM_005270412.3 | XP_005270469.1 | 7 | 1407 | |||
XM_005270413.4 | XP_005270470.1 | 7 | 1684 | |||
XM_011540569.2 | XP_011538871.1 | 6 | 1540 | |||
GenBank mRNA | AK024011.1 | - | 7 | 945 | ||
AK093320.1 | - | 7 | 1219 | |||
AK293704.1 | - | 5 | 798 | |||
AY169960.1 | - | 7 | 1421 | |||
BC009364.2 | - | 7 | 858 | 4302712 | ||
CR457320.1 | - | 7 | 841 |