Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031908.4 | NP_114114.2 | 3 | 1829 | ||
XM_011534425.2 | XP_011532727.1 | 4 | 1961 | |||
XM_011534427.2 | XP_011532729.1 | 5 | 1852 | |||
GenBank mRNA | BC011699.2 | - | 3 | 1810 |