Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031910.3 | NP_114116.3 | 2 | 347 | ||
NM_182486.1 | NP_872292.1 | 2 | 347 | |||
XM_011529857.1 | XP_011528159.1 | 3 | 3204 | |||
XM_017028569.1 | XP_016884058.1 | 2 | 2496 | |||
XM_017028570.1 | XP_016884059.1 | 4 | 1114 | |||
XM_017028571.1 | XP_016884060.1 | 4 | 2852 | |||
XM_017028572.1 | XP_016884061.1 | 4 | 2841 | |||
XM_017028573.1 | XP_016884062.1 | 4 | 630 | |||
XM_017028574.1 | XP_016884063.1 | 3 | 423 | |||
GenBank mRNA | AF329842.1 | - | 2 | 285 | ||
AK128125.1 | - | 6 | 3760 | |||
AK226149.1 | - | 1 | 870 | |||
AK314060.1 | - | 2 | 304 | |||
AY358374.1 | - | 4 | 484 | |||
BC020551.1 | - | 2 | 346 | 2900683 | ||
BU147265.1 | - | 2 | 349 | |||
BU535692.1 | - | 1 | 86 | |||
CB961508.1 | - | 2 | 331 | |||
CR456451.1 | - | 2 | 297 |