Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324113.1 | NP_001311042.1 | 11 | 1913 | ||
NM_001324114.1 | NP_001311043.1 | 10 | 1801 | |||
NM_001324115.1 | NP_001311044.1 | 11 | 2140 | |||
NM_001324116.1 | NP_001311045.1 | 11 | 2079 | |||
NM_001324117.1 | NP_001311046.1 | 11 | 2076 | |||
NM_003115.4 | NP_003106.3 | 10 | 1962 | |||
XM_011509913.2 | XP_011508215.1 | 12 | 1912 | |||
XM_011509914.2 | XP_011508216.1 | 12 | 1964 | |||
GenBank mRNA | BC009377.2 | - | 10 | 1935 | 4121439 | |
CB125476.1 | - | 1 | 76 | |||
S73498.1 | - | 10 | 1865 |