Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135170.1 | NP_001128642.1 | 3 | 1072 | ||
NM_001135171.1 | NP_001128643.1 | 3 | 1019 | |||
NM_031911.4 | NP_114117.1 | 3 | 902 | |||
XM_011513772.1 | XP_011512074.1 | 4 | 942 | |||
XM_011513773.1 | XP_011512075.1 | 4 | 991 | |||
GenBank mRNA | AF329839.1 | - | 3 | 877 | ||
AK314802.1 | - | 3 | 1020 | |||
BC022187.2 | - | 3 | 1020 | 4825040 | ||
BC024015.2 | - | 3 | 1020 | 4823626 | ||
BX647781.1 | - | 3 | 1072 |