Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTYH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032646.5 | NP_116035.5 | 13 | 1527 | ||
NM_052869.1 | NP_443101.1 | 7 | 1337 | |||
XM_005257824.3 | XP_005257881.1 | 13 | 1455 | |||
GenBank mRNA | AF319952.1 | - | 13 | 1506 | ||
AK058197.1 | - | 2 | 227 | |||
AK091085.1 | - | 11 | 1208 | |||
AK126955.1 | - | 7 | 1745 | |||
AK296511.1 | - | 13 | 1450 | |||
AK315163.1 | - | 13 | 1513 | |||
AL831858.1 | - | 3 | 273 | |||
BC005168.1 | - | 13 | 1502 | |||
BC107492.1 | - | 13 | 1523 | 40033786 | ||
BC107602.1 | - | 13 | 1523 | 40033791 | ||
BX647912.1 | - | 7 | 1337 |