Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032796.3 | NP_116185.2 | 6 | 748 | ||
NR_033181.1 | 6 | 710 | ||||
GenBank mRNA | AF168955.1 | - | 6 | 700 | ||
AF305828.1 | - | 3 | 240 | |||
AF338728.1 | - | 6 | 700 | |||
AK027401.1 | - | 6 | 690 | |||
AK126173.1 | - | 1 | 1663 | |||
AK222856.1 | - | 6 | 682 | |||
AK295322.1 | - | 6 | 685 | |||
AK303783.1 | - | 6 | 664 | |||
AK314675.1 | - | 6 | 720 | |||
BC001468.1 | - | 6 | 679 | |||
BC014657.1 | - | 6 | 668 | 4640905 | ||
CR749849.1 | - | 6 | 734 |