Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174044.1 | NP_001167515.1 | 5 | 1779 | ||
NM_001174045.1 | NP_001167516.1 | 6 | 1774 | |||
NM_001174046.1 | NP_001167517.1 | 5 | 1689 | |||
NM_003117.4 | NP_003108.2 | 5 | 1600 | |||
NM_153189.2 | NP_694859.1 | 5 | 1600 | |||
GenBank mRNA | AK292229.1 | - | 5 | 1689 | ||
AY920278.1 | - | 5 | 1775 | |||
AY920279.1 | - | 5 | 1596 | |||
AY920280.1 | - | 6 | 1749 | |||
AY920281.1 | - | 5 | 1664 | |||
BC026163.1 | - | 5 | 1523 | 4838230 | ||
EU794686.1 | - | 5 | 1563 | |||
L13781.1 | - | 3 | 1262 | |||
S67798.1 | - | 5 | 1534 | |||
X84347.1 | - | 4 | 1457 |