Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53INP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135733.1 | NP_001129205.1 | 3 | 711 | ||
NM_033285.3 | NP_150601.1 | 3 | 711 | |||
XM_011517386.2 | XP_011515688.1 | 3 | 563 | |||
GenBank mRNA | AB017926.1 | - | 2 | 366 | ||
AB017927.1 | - | 2 | 366 | |||
AF409114.1 | - | 3 | 683 | |||
AF409115.1 | - | 3 | 683 | |||
AK125880.1 | - | 3 | 690 | |||
AK315071.1 | - | 3 | 653 | |||
AL833652.1 | - | 2 | 291 | |||
BC074813.2 | - | 2 | 359 | 30915317 | ||
BC074868.2 | - | 2 | 359 | 30915467 |