Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53INP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135733.1 | NP_001129205.1 | 5 | 1102 | ||
NM_033285.3 | NP_150601.1 | 4 | 1064 | |||
XM_011517386.2 | XP_011515688.1 | 4 | 916 | |||
GenBank mRNA | AB017926.1 | - | 3 | 719 | ||
AB017927.1 | - | 4 | 757 | |||
AF409114.1 | - | 5 | 1074 | |||
AF409115.1 | - | 4 | 1036 | |||
AK125880.1 | - | 5 | 1081 | |||
AK315071.1 | - | 4 | 1006 | |||
AL833652.1 | - | 4 | 682 | |||
BC074813.2 | - | 3 | 712 | 30915317 | ||
BC074868.2 | - | 3 | 712 | 30915467 |