Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033515.2 | NP_277050.2 | 5 | 815 | ||
XM_005267212.2 | XP_005267269.2 | 5 | 891 | |||
XM_005267213.1 | XP_005267270.1 | 5 | 815 | |||
XM_017011494.1 | XP_016866983.1 | 5 | 891 | |||
GenBank mRNA | AB053293.1 | - | 4 | 613 | ||
AK098594.1 | - | 5 | 815 | |||
AK307796.1 | - | 7 | 891 | |||
AL834511.1 | - | 5 | 753 | |||
AY643712.1 | - | 5 | 816 | |||
BC017223.2 | - | 5 | 737 | |||
BC039611.1 | - | 5 | 744 | |||
BC052295.1 | - | 2 | 153 | |||
BC062417.1 | - | 5 | 724 | |||
BC101708.1 | - | 5 | 809 | 8069214 | ||
BC107416.1 | - | 5 | 771 | 4040931 | ||
BC111940.1 | - | 5 | 809 | 8327408 | ||
BC143531.1 | - | 5 | 809 | |||
BM557223.1 | - | 5 | 752 |