Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052874.4 | NP_443106.1 | 4, 5 | 530 | ||
XM_017022893.1 | XP_016878382.1 | 4, 5 | 285 | |||
GenBank mRNA | AY028792.1 | - | 4, 5 | 346 | ||
AY995211.1 | - | 4, 5 | 288 | |||
BC038359.1 | - | 1, 2 | 36 | |||
BC062298.1 | - | 4, 5 | 342 | 6375788 | ||
DN990202.1 | - | 4, 5 | 524 |