Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052885.3 | NP_443117.3 | 3 | 844 | ||
XM_011537847.2 | XP_011536149.1 | 3 | 1070 | |||
XM_011537849.2 | XP_011536151.1 | 3 | 1070 | |||
XM_011537850.2 | XP_011536152.1 | 3 | 1070 | |||
XM_017018764.1 | XP_016874253.1 | 3 | 317 | |||
XM_017018765.1 | XP_016874254.1 | 3 | 955 | |||
XM_017018766.1 | XP_016874255.1 | 2 | 102 | |||
GenBank mRNA | AJ315644.1 | - | 3 | 844 | ||
BC047507.1 | - | 3 | 1074 | 5272386 | ||
BC117117.1 | - | 3 | 788 | 40125668 | ||
BC117119.1 | - | 3 | 788 | 40125670 |