Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030288.2 | NP_001025459.1 | 2 | 839 | ||
NM_003123.4 | NP_003114.1 | 2 | 806 | |||
GenBank mRNA | AK292626.1 | - | 2 | 778 | ||
AK313750.1 | - | 2 | 778 | |||
BC012350.1 | - | 2 | 726 | 4564347 | ||
J04536.1 | - | 2 | 766 | |||
X60502.1 | - | 2 | 810 |