Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052897.3 | NP_443129.3 | 13 | 3761 | ||
XM_005268617.1 | XP_005268674.1 | 13 | 3817 | |||
XM_005268618.1 | XP_005268675.1 | 13 | 3913 | |||
XM_006719217.1 | XP_006719280.1 | 13 | 3662 | |||
XM_006719218.1 | XP_006719281.1 | 13 | 3650 | |||
XM_011537851.2 | XP_011536153.1 | 13 | 4650 | |||
GenBank mRNA | AB067474.1 | - | 10 | 3430 | ||
BC065530.1 | - | 13 | 3761 | 6152748 | ||
CR627478.1 | - | 6 | 1341 |