Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006641.3 | NP_001006642.1 | 2 | 402 | ||
NM_001006642.3 | NP_001006643.1 | 2 | 647 | |||
NM_001265614.2 | NP_001252543.1 | 2 | 647 | |||
NM_052901.4 | NP_443133.2 | 2 | 265 | |||
NR_049766.2 | 2 | 308 | ||||
XM_005251688.1 | XP_005251745.1 | 2 | 402 | |||
XM_005251689.4 | XP_005251746.1 | 2 | 661 | |||
XM_006716948.2 | XP_006717011.1 | 2 | 803 | |||
XM_006716949.3 | XP_006717012.1 | 2 | 663 | |||
GenBank mRNA | AB067483.1 | - | 2 | 646 | ||
AJ619963.1 | - | 2 | 566 | |||
AJ619989.1 | - | 2 | 190 | |||
AJ619990.1 | - | 2 | 285 | |||
AJ619991.1 | - | 2 | 247 | |||
AK290705.1 | - | 2 | 238 | |||
AK290991.1 | - | 2 | 402 | |||
AL559911.3 | - | 2 | 409 | |||
AY358515.1 | - | 2 | 240 | |||
BC089448.1 | - | 2 | 256 | 30558551 | ||
BC103930.1 | - | 2 | 207 | 40008034 | ||
BC103931.1 | - | 2 | 207 | 40008035 | ||
BC103932.1 | - | 2 | 207 | 40008036 | ||
BC103933.1 | - | 2 | 207 | 40008037 | ||
BQ637100.1 | - | 2 | 265 |