Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052905.3 | NP_443137.2 | 9 | 1207 | ||
XM_005246263.2 | XP_005246320.1 | 9 | 1274 | |||
XM_005246265.3 | XP_005246322.1 | 9 | 1274 | |||
XM_011510530.1 | XP_011508832.1 | 9 | 1274 | |||
XM_011510531.1 | XP_011508833.1 | 9 | 1274 | |||
XM_011510532.2 | XP_011508834.1 | 9 | 1274 | |||
XM_011510533.2 | XP_011508835.1 | 9 | 1274 | |||
XM_011510534.1 | XP_011508836.1 | 9 | 1274 | |||
XM_011510535.2 | XP_011508837.1 | 8 | 1121 | |||
XM_011510536.2 | XP_011508838.1 | 3 | 225 | |||
GenBank mRNA | AB067489.1 | - | 9 | 975 | ||
AL834396.1 | - | 7 | 588 | |||
BC167159.1 | - | 9 | 975 | 8862581 |