Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM200A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258276.1 | NP_001245205.1 | 2 | 1293 | ||
NM_001258277.1 | NP_001245206.1 | 3 | 1357 | |||
NM_001258278.1 | NP_001245207.1 | 2 | 1312 | |||
NM_052913.2 | NP_443145.1 | 2 | 3411 | |||
XM_005266816.2 | XP_005266873.1 | 4 | 1436 | |||
XM_011535419.2 | XP_011533721.1 | 2 | 1113 | |||
XM_017010230.1 | XP_016865719.1 | 2 | 2088 | |||
GenBank mRNA | AB052099.1 | - | 2 | 3411 | ||
AB067500.1 | - | 3 | 1904 | |||
AK290166.1 | - | 2 | 1306 | |||
AK294359.1 | - | 1 | 736 | |||
AK315824.1 | - | 2 | 1189 | |||
BC044246.1 | - | 3 | 1176 | 5760073 |