Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134486.2 | NP_001127958.1 | 2 | 546 | ||
NM_052942.3 | NP_443174.1 | 3 | 654 | |||
GenBank mRNA | AF288815.1 | - | 1 | 117 | ||
AF328727.1 | - | 3 | 617 | |||
AF430642.1 | - | 3 | 655 | |||
AF430643.1 | - | 2 | 535 | |||
AK090479.1 | - | 1 | 3366 | |||
AK315064.1 | - | 3 | 265 | |||
AY358953.1 | - | 2 | 480 | |||
BC031639.1 | - | 3 | 641 | 5173241 |