Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052950.3 | NP_443182.1 | 8 | 1104 | ||
XM_011534914.1 | XP_011533216.1 | 8 | 995 | |||
XM_011534915.2 | XP_011533217.1 | 7 | 1184 | |||
XM_017020386.1 | XP_016875875.1 | 8 | 6908 | |||
GenBank mRNA | AF411978.1 | - | 8 | 764 | ||
AK058149.1 | - | 6 | 550 | |||
AK309815.1 | - | 8 | 957 | |||
AK311177.1 | - | 6 | 762 | |||
BC014004.2 | - | 8 | 944 | 3842589 |