Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318890.1 | NP_001305819.1 | 14 | 1888 | ||
NM_052956.2 | NP_443188.2 | 13 | 1752 | |||
NR_134918.1 | 11 | 1420 | ||||
XM_006721016.2 | XP_006721079.1 | 12 | 1629 | |||
XM_006721017.2 | XP_006721080.1 | 11 | 1548 | |||
XM_006721018.2 | XP_006721081.1 | 11 | 1110 | |||
XM_011545730.2 | XP_011544032.1 | 9 | 1257 | |||
XM_011545731.2 | XP_011544033.1 | 11 | 1927 | |||
XM_011545732.2 | XP_011544034.1 | 12 | 1169 | |||
XM_017022915.1 | XP_016878404.1 | 12 | 2579 | |||
GenBank mRNA | AB059429.1 | - | 13 | 1752 | ||
AK092574.1 | - | 10 | 1540 | |||
AK314282.1 | - | 14 | 1844 | |||
BC125177.1 | - | 11 | 1420 | |||
BC125178.1 | - | 13 | 1752 | 40121956 |