Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSEN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052965.3 | NP_443197.1 | 4, 5 | 602 | ||
NR_023349.2 | 5, 6 | 719 | ||||
NR_125335.1 | 5, 6 | 682 | ||||
GenBank mRNA | AF288394.1 | - | 4, 5 | 577 | ||
AK098196.1 | - | 4, 5 | 500 | |||
AK307241.1 | - | 5, 6 | 610 | |||
BC022030.1 | - | 4, 5 | 547 | 4824025 | ||
BU195337.1 | - | 5, 6 | 654 |