Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052967.1 | NP_443199.1 | 1 | 175 | ||
GenBank mRNA | BC069127.1 | - | 1 | 175 | 7216983 | |
BC069345.1 | - | 1 | 175 | 7262308 | ||
BC101176.2 | - | 1 | 175 | 40016384 | ||
BC101177.1 | - | 1 | 175 | 40016387 |
Set Membership: |
G-protein coupled receptor |