Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300854.1 | NP_001287783.1 | 6 | 629 | ||
NM_053283.3 | NP_444513.1 | 5 | 565 | |||
GenBank mRNA | AF144011.1 | - | 5 | 435 | ||
BC062682.1 | - | 5 | 437 | 30369929 | ||
BC069108.1 | - | 5 | 435 | 7216925 | ||
EF503687.1 | - | 5 | 565 | |||
EF503688.1 | - | 6 | 732 | |||
EF503689.1 | - | 6 | 629 |