Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057161.3 | NP_476502.1 | 9 | 1279 | ||
NR_040101.1 | 8 | 1163 | ||||
GenBank mRNA | AB055925.1 | - | 9 | 1143 | ||
AK097719.1 | - | 7 | 1005 | |||
AK290384.1 | - | 9 | 1135 | |||
AK302336.1 | - | 8 | 918 | |||
AK308917.1 | - | 8 | 1285 | |||
BC000295.2 | - | 9 | 1099 | 2819570 | ||
BC001789.1 | - | 9 | 1114 | 3355470 | ||
BC001793.2 | - | 9 | 1077 | 3352397 | ||
BC007296.1 | - | 9 | 1129 | 3349731 | ||
BC009460.1 | - | 9 | 1127 | 3537223 | ||
BC021546.1 | - | 9 | 1116 | 3351635 | ||
BC041793.1 | - | 8 | 1017 | 5288540 | ||
BC045612.1 | - | 8 | 1017 | 5313255 |