Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080476.4 | NP_536724.1 | 12 | 1790 | ||
XM_011528542.2 | XP_011526844.1 | 6 | 1081 | |||
XM_017027664.1 | XP_016883153.1 | 11 | 1464 | |||
GenBank mRNA | AK075507.1 | - | 12 | 1615 | ||
AK095044.1 | - | 1 | 3085 | |||
AK095356.1 | - | 1 | 3775 | |||
AY358816.1 | - | 12 | 1620 | |||
AY422169.1 | - | 12 | 1790 | |||
BC030512.1 | - | 12 | 1601 | 4942673 |