Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256798.1 | NP_001243727.1 | 11 | 2335 | ||
NM_080616.4 | NP_542183.2 | 8 | 1674 | |||
GenBank mRNA | AK056286.1 | - | 8 | 1477 |