Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289112.1 | NP_001276041.1 | 16 | 2184 | ||
NM_130808.2 | NP_570720.1 | 16 | 2812 | |||
NM_153429.1 | NP_702907.1 | 17 | 2172 | |||
XM_011512406.2 | XP_011510708.1 | 15 | 1959 | |||
XM_011512407.2 | XP_011510709.1 | 15 | 2216 | |||
XM_011512408.2 | XP_011510710.1 | 15 | 3344 | |||
XM_017005693.1 | XP_016861182.1 | 19 | 3565 | |||
XM_017005695.1 | XP_016861184.1 | 18 | 2435 | |||
XM_017005696.1 | XP_016861185.1 | 10 | 1329 | |||
GenBank mRNA | AF465771.1 | - | 17 | 2173 | ||
AK056175.1 | - | 16 | 2284 | |||
AK128117.1 | - | 20 | 4062 | |||
BC014396.1 | - | 16 | 2370 | 3162811 | ||
BC028613.1 | - | 3 | 938 | |||
BM930746.1 | - | 2 | 509 |