Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135057.2 | NP_001128529.2 | 3 | 1042 | ||
NM_130830.4 | NP_570843.2 | 2 | 985 | |||
GenBank mRNA | AB071037.1 | - | 1 | 899 | ||
AK125112.1 | - | 2 | 985 | |||
BC101064.1 | - | 3 | 1030 | 40008605 | ||
BC101065.1 | - | 2 | 973 | 40008610 |