Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270614.1 | NP_001257543.1 | 5 | 1737 | ||
NM_001270615.1 | NP_001257544.1 | 5 | 1804 | |||
NM_133328.3 | NP_579874.1 | 5 | 1819 | |||
NR_073046.1 | 5 | 1551 | ||||
NR_073047.1 | 4 | 1371 | ||||
NR_073048.1 | 4 | 1453 | ||||
NR_073049.1 | 5 | 1475 | ||||
NR_073050.1 | 4 | 1438 | ||||
NR_073051.1 | 5 | 1537 | ||||
NR_073052.1 | 3 | 1312 | ||||
XM_011526569.1 | XP_011524871.1 | 7 | 1863 | |||
XM_011526571.1 | XP_011524873.1 | 6 | 1841 | |||
XM_011526572.1 | XP_011524874.1 | 6 | 1765 | |||
XM_017026402.1 | XP_016881891.1 | 7 | 1857 | |||
XM_017026403.1 | XP_016881892.1 | 7 | 1925 | |||
GenBank mRNA | AK075328.1 | - | 6 | 1684 | ||
AK130203.1 | - | 5 | 1459 | |||
AY125488.1 | - | 5 | 1688 | |||
BC013372.2 | - | 5 | 1681 | 3941907 | ||
BC027930.1 | - | 5 | 1688 | 5199772 |