Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133373.4 | NP_588614.1 | 14, 15 | 2408 | ||
GenBank mRNA | AB075844.1 | - | 14, 15 | 2190 | ||
AK074240.1 | - | 14, 15 | 2389 | |||
AL834392.1 | - | 14, 15 | 2250 | |||
BC010668.1 | - | 12, 13 | 1770 | |||
BC072384.1 | - | 14, 15 | 2373 | 6378601 |