Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIN3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133445.2 | NP_597702.2 | 2 | 1767 | ||
XM_011518211.2 | XP_011516513.1 | 2 | 1191 | |||
GenBank mRNA | AB075853.1 | - | 2 | 1767 | ||
AF416558.1 | - | 2 | 1179 | |||
AJ416950.2 | - | 2 | 1181 | |||
AK127342.1 | - | 2 | 734 | |||
BC132866.1 | - | 2 | 1593 | 40146888 | ||
BC132868.1 | - | 2 | 1593 | 40146890 | ||
BC167432.1 | - | 2 | 1767 | 8860109 |