Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159587.2 | NP_001153059.1 | 8 | 913 | ||
NM_001159588.2 | NP_001153060.1 | 7 | 682 | |||
NM_133493.4 | NP_598000.2 | 8 | 913 | |||
XM_005248659.3 | XP_005248716.1 | 9 | 1060 | |||
XM_011535473.1 | XP_011533775.1 | 8 | 758 | |||
XM_017010283.1 | XP_016865772.1 | 6 | 525 | |||
GenBank mRNA | AF410459.1 | - | 8 | 894 | ||
AK095888.1 | - | 8 | 847 | |||
AK313636.1 | - | 8 | 893 | |||
AY149920.1 | - | 8 | 782 | |||
AY788891.1 | - | 8 | 807 | |||
BX641095.1 | - | 7 | 569 | |||
EF553520.1 | - | 8 | 812 |