Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134268.4 | NP_599030.1 | 2, 3 | 732 | ||
XM_005257005.2 | XP_005257062.1 | 2, 3 | 379 | |||
XM_017024116.1 | XP_016879605.1 | 3, 4 | 597 | |||
GenBank mRNA | AB057769.1 | - | 2, 3 | 412 | ||
AJ315162.1 | - | 2, 3 | 369 | |||
AK098057.1 | - | 2, 3 | 379 | |||
BC029798.1 | - | 2, 3 | 408 | 5193583 | ||
BQ881744.1 | - | 2, 3 | 732 |