Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012426.1 | NP_001012426.1 | 15 | 2163 | ||
NM_001012427.1 | NP_001012427.1 | 15 | 2157 | |||
NM_138457.2 | NP_612466.1 | 15 | 2124 | |||
XM_006714991.3 | XP_006715054.1 | 15 | 1965 | |||
XM_011514289.2 | XP_011512591.1 | 15 | 2061 | |||
XM_011514290.2 | XP_011512592.1 | 15 | 2056 | |||
XM_011514291.2 | XP_011512593.1 | 15 | 2024 | |||
XM_011514292.2 | XP_011512594.1 | 14 | 1966 | |||
XM_011514293.2 | XP_011512595.1 | 13 | 1582 | |||
XM_017010233.1 | XP_016865722.1 | 15 | 3193 | |||
XM_017010234.1 | XP_016865723.1 | 15 | 2061 | |||
GenBank mRNA | AB080747.1 | - | 14 | 1717 | ||
AL832437.1 | - | 4 | 323 | |||
BC013030.2 | - | 2 | 126 | |||
BC016874.1 | - | 9 | 903 | |||
BC034931.1 | - | 5 | 551 | |||
BC040962.1 | - | 15 | 2124 | 5527139 | ||
BC052803.1 | - | 15 | 1935 | 6420057 |