Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003179.2 | NP_003170.1 | 7 | 1428 | ||
GenBank mRNA | BC032385.1 | - | 1 | 169 | ||
BC064550.1 | - | 7 | 1428 | 5768012 |