Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYND19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138462.2 | NP_612471.1 | 5 | 724 | ||
XM_005266052.4 | XP_005266109.1 | 5 | 1153 | |||
GenBank mRNA | AJ298882.2 | - | 5 | 732 | ||
AX766496.1 | - | 5 | 732 | |||
BC012948.1 | - | 5 | 687 | 4519078 |