Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080722.4 | NP_542453.2 | 22 | 4003 | ||
NM_139155.3 | NP_631894.2 | 22 | 4012 | |||
XM_011539300.2 | XP_011537602.1 | 19 | 3486 | |||
XM_011539301.2 | XP_011537603.1 | 19 | 3009 | |||
XM_011539302.2 | XP_011537604.1 | 19 | 2886 | |||
XM_011539309.1 | XP_011537611.1 | 15 | 2596 | |||
GenBank mRNA | AJ345098.1 | - | 22 | 3706 |