Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALS2CR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127391.2 | NP_001120863.1 | 15 | 2001 | ||
NM_001289993.1 | NP_001276922.1 | 15 | 1669 | |||
NM_139163.3 | NP_631902.2 | 15 | 2070 | |||
NR_110620.1 | 14 | 2167 | ||||
XM_011510606.2 | XP_011508908.1 | 15 | 2016 | |||
XM_011510610.2 | XP_011508912.1 | 13 | 1461 | |||
XM_011510612.2 | XP_011508914.1 | 9 | 1168 | |||
XM_017003362.1 | XP_016858851.1 | 13 | 1502 | |||
GenBank mRNA | AB053314.1 | - | 14 | 1655 |