Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGCZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322879.1 | NP_001309808.1 | 7 | 1597 | ||
NM_001322880.1 | NP_001309809.1 | 7 | 1576 | |||
NM_001322881.1 | NP_001309810.1 | 7 | 1482 | |||
NM_139167.3 | NP_631906.2 | 8 | 1699 | |||
GenBank mRNA | AY028700.1 | - | 7 | 1000 |