Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127255.1 | NP_001120727.1 | 4 | 584 | ||
NM_139176.3 | NP_631915.2 | 4 | 584 | |||
NM_206828.3 | NP_996611.2 | 4 | 584 | |||
XM_006723075.3 | XP_006723138.1 | 5 | 819 | |||
XM_006723076.3 | XP_006723139.1 | 4 | 782 | |||
XM_011526596.2 | XP_011524898.1 | 6 | 1026 | |||
XM_011526599.2 | XP_011524901.1 | 6 | 1037 | |||
XM_011526601.2 | XP_011524903.1 | 6 | 1026 | |||
GenBank mRNA | AF464765.1 | - | 3 | 554 | ||
AK302655.1 | - | 6 | 995 | |||
AK313964.1 | - | 6 | 979 | |||
AY154462.1 | - | 4 | 578 | |||
BC109124.2 | - | 4 | 576 | 40036025 | ||
BC109125.1 | - | 4 | 576 | 40036028 |