Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127715.2 | NP_001121187.1 | 5, 6 | 739 | ||
NM_139244.4 | NP_640337.3 | 5, 6 | 739 | |||
XM_006715339.2 | XP_006715402.1 | 5, 6 | 913 | |||
XM_011535453.1 | XP_011533755.1 | 4, 5 | 416 | |||
XM_017010268.1 | XP_016865757.1 | 5, 6 | 776 | |||
GenBank mRNA | AK128121.1 | - | 4, 5 | 411 | ||
AL834152.1 | - | 5, 6 | 725 | |||
BC107737.1 | - | 5, 6 | 676 | |||
BC113382.1 | - | 5, 6 | 612 | 8322434 | ||
BC113408.1 | - | 5, 6 | 612 | 8322460 |