Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003192.2 | NP_003183.1 | 1 | 1289 | ||
GenBank mRNA | BC017479.1 | - | 2 | 1780 | 3357283 | |
BC020170.1 | - | 1 | 1229 | 3536688 | ||
U61234.1 | - | 1 | 1239 |