Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139278.2 | NP_644807.1 | 8 | 1792 | ||
GenBank mRNA | AF467956.1 | - | 8 | 1713 | ||
AJ487518.1 | - | 8 | 1922 | |||
AJ487961.1 | - | 8 | 1792 | |||
AK090946.1 | - | 5 | 1113 | |||
AY358116.1 | - | 8 | 1530 | |||
BC042542.1 | - | 2 | 821 | |||
BC142997.1 | - | 7 | 1640 | 8860454 |