Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORMDL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320801.1 | NP_001307730.1 | 6 | 1937 | ||
NM_001320802.1 | NP_001307731.1 | 4 | 775 | |||
NM_001320803.1 | NP_001307732.1 | 4 | 489 | |||
NM_139280.3 | NP_644809.1 | 4 | 780 | |||
XM_017025345.1 | XP_016880834.1 | 4 | 633 | |||
XM_017025346.1 | XP_016880835.1 | 4 | 546 | |||
XM_017025347.1 | XP_016880836.1 | 4 | 726 | |||
XM_017025348.1 | XP_016880837.1 | 4 | 472 | |||
GenBank mRNA | AF373101.1 | - | 4 | 516 | ||
AF373102.1 | - | 4 | 586 | |||
AF395708.1 | - | 4 | 571 | |||
AK074811.1 | - | 4 | 577 | |||
AK075212.1 | - | 4 | 532 | |||
AK093063.1 | - | 6 | 1932 | |||
AX775843.1 | - | 4 | 534 | |||
AY357943.1 | - | 3 | 389 | |||
BC017087.1 | - | 4 | 546 | 3850211 | ||
BC071833.1 | - | 4 | 525 | 6340437 | ||
DB042662.1 | - | 4 | 490 |