Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORMDL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320801.1 | NP_001307730.1 | 6 | 3449 | ||
NM_001320802.1 | NP_001307731.1 | 4 | 2287 | |||
NM_001320803.1 | NP_001307732.1 | 4 | 2001 | |||
NM_139280.3 | NP_644809.1 | 4 | 2292 | |||
XM_017025345.1 | XP_016880834.1 | 4 | 2145 | |||
XM_017025346.1 | XP_016880835.1 | 4 | 2058 | |||
XM_017025347.1 | XP_016880836.1 | 4 | 2238 | |||
XM_017025348.1 | XP_016880837.1 | 4 | 1984 | |||
GenBank mRNA | AF151043.1 | - | 1 | 731 | ||
AF373101.1 | - | 4 | 2028 | |||
AK074811.1 | - | 4 | 2089 | |||
AK075212.1 | - | 4 | 2044 | |||
AX775843.1 | - | 4 | 2046 | |||
BC017087.1 | - | 4 | 2058 | 3850211 | ||
BC018837.1 | - | 1 | 762 | |||
BC071833.1 | - | 4 | 2037 | 6340437 |