Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144601.3 | NP_653202.1 | 6 | 1539 | ||
NM_181553.2 | NP_853531.1 | 5 | 1375 | |||
NR_037613.1 | 6 | 1759 | ||||
NR_037619.1 | 5 | 1432 | ||||
XM_006721131.1 | XP_006721194.1 | 6 | 1583 | |||
XM_011522861.1 | XP_011521163.1 | 4 | 1212 | |||
GenBank mRNA | AF479813.1 | - | 6 | 1513 | ||
AK056324.1 | - | 6 | 1759 | |||
AK075075.1 | - | 4 | 1072 | |||
AY166714.1 | - | 2 | 458 | |||
BC023509.1 | - | 5 | 1281 | 3960441 | ||
BC023591.2 | - | 5 | 1273 | 4638047 | ||
BC036042.1 | - | 5 | 1335 | 5271808 | ||
BC040227.1 | - | 5 | 1326 | 5187826 | ||
BC047701.1 | - | 5 | 1375 | 5770132 |